MilliporeSigma
  • Vitamin E prevents apoptosis in cortical neurons during hypoxia and oxygen reperfusion.
Vitamin E prevents apoptosis in cortical neurons during hypoxia and oxygen reperfusion.
Laboratory investigation; a journal of technical methods and pathology (1998-12-05)
M Tagami, K Yamagata, K Ikeda, Y Nara, H Fujino, A Kubota, F Numano, Y Yamori
ABSTRACT
Cerebral ischemia followed by oxygen reperfusion induces apoptosis in hippocampal neurons in stroke-prone spontaneously hypertensive rats (SHRSP) but not in Wistar Kyoto rats (WKY). The overproduction of oxygen-free radicals that occurs in the tissues of SHRSP is implicated in reoxygenation injury after hypoxia. Antioxidants inhibit reoxygenation injury in hippocampal slices, and temporal cortices in Alzheimer's disease increase sensitivity to oxygen-free radicals. Because this sensitivity may contribute to the development of the disease, we have studied hypoxia and oxygen reperfusion using cortical neurons isolated from WKY and SHRSP (at 15 days of gestation). We have tried to determine whether cortical neurons are damaged under these conditions, and whether neurons from SHRSP are more vulnerable than those from WKY. We have tried also to verify whether neuronal damage is minimized by vitamin E using the following techniques: (a) Trypan blue staining, (b) in situ staining of apoptosis, (c) ultrastructural examination, and (d) measurement of lactic dehydrogenase (LDH) activity in the bathing medium. Furthermore, we have examined the mechanisms involved in the development of neuronal damage and have studied ways of minimizing it. We demonstrated that 36 hours of hypoxia significantly increased the rate of cell death in SHRSP (p < 0.01), although 12 to 24 hours of hypoxia did not increase cell death in either WKY or SHRSP. In addition, 6 to 36 hours of hypoxia and 1.5 to 5 hours of oxygen reperfusion heavily damaged cells of both WKY and SHRSP, and most became apoptotic or necrotic. In contrast, cells incubated with 50 to 300 microg/ml of vitamin E remained intact, although 10 to 20 microg/ml of vitamin E did not totally preserve the cells. Moreover, vitamin E protected the neurons from high concentrations of sodium nitroprusside (nitric oxide donor) in a dose-dependent manner. Vitamin E, when added to the cells, increased in concentration in a time-dependent manner over a 24-hour period and in a dose-dependent manner below 200 microg/ml, and it was detected mostly in the mitochondria. We also demonstrated that serial treatments with allopurinol (a xanthine oxidase inhibitor) or superoxide dismutase preserved neurons during hypoxia and oxygen reperfusion. These data indicate that SHRSP neurons are weaker than WKY neurons in long-term hypoxia; oxygen radical generation occurs in the early minutes after reperfusion, and then the oxygen-free radicals cause heavy damage to the cells; and antioxidants including vitamin E react with the radicals, thereby preventing apoptosis and necrosis. Therefore, antioxidants appear to be the most important agents in lowering oxygen-free radical damage in cortical neurons.
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Pricing and availability is not currently available.