MilliporeSigma
  • The discovery and development of inhibitors of fatty acid amide hydrolase (FAAH).
The discovery and development of inhibitors of fatty acid amide hydrolase (FAAH).
Bioorganic & medicinal chemistry letters (2011-07-19)
Katerina Otrubova, Cyrine Ezzili, Dale L Boger
ABSTRACT
A summary of the discovery and advancement of inhibitors of fatty acid amide hydrolase (FAAH) is presented.
Sigma-Aldrich
1-Naphthyl-N-methylcarbamate, 97%