MilliporeSigma
  • Furanocoumarin with Phytotoxic Activity from the Leaves of Amyris elemifera (Rutaceae).
Furanocoumarin with Phytotoxic Activity from the Leaves of Amyris elemifera (Rutaceae).
ACS omega (2021-01-19)
Kumudini M Meepagala, Amy K Bracken, Frank R Fronczek, Robert D Johnson, David E Wedge, Stephen O Duke
ABSTRACT
Bioassay-guided fractionation of the ethyl acetate extract of Amyris elemifera leaves was carried out to identify phytotoxic and antifungal constituents. A novel phytotoxic furanocoumarin 8-(3-methylbut-2-enyloxy)-marmesin acetate (1) and its deacyl analog 8-(3-methylbut-2-enyloxy)-marmesin (2) were isolated. The X-ray crystal structure determination is reported for the first time for 1. Both 1 and 2 have the S configuration at C-2' based on X-ray crystallographic data. Both these compounds inhibited the growth of the dicot Lactuca sativa (lettuce) and the monocot Agrostis stolonifera with a more pronounced inhibitory effect on the monocots at 330 μM by 1. In Lemna paucicostata Hegelm phytotoxicity bioassay, the IC50 value for 1 was 26 μM, whereas 2 had an IC50 value of 102 μM. Compounds 1 and 2 were weakly antifungal against Colletotrichum fragariae Brooks in TLC bioautography.
Sigma-Aldrich
Hoagland′s No. 2 Basal Salt Mixture, powder, suitable for plant cell culture