MilliporeSigma
  • In vitro potentiation of carbapenems with ME1071, a novel metallo-beta-lactamase inhibitor, against metallo-beta-lactamase- producing Pseudomonas aeruginosa clinical isolates.
In vitro potentiation of carbapenems with ME1071, a novel metallo-beta-lactamase inhibitor, against metallo-beta-lactamase- producing Pseudomonas aeruginosa clinical isolates.
Antimicrobial agents and chemotherapy (2010-07-08)
Yoshikazu Ishii, Maki Eto, Yoko Mano, Kazuhiro Tateda, Keizo Yamaguchi
ABSTRACT
ME1071, a maleic acid derivative, is a novel specific inhibitor for metallo-beta-lactamases (MBL). In this study, the potentiation of ME1071 in combination with several beta-lactams was evaluated using MBL-producing Pseudomonas aeruginosa isolates. The rates of susceptibility of MBL producers to carbapenems (imipenem, biapenem, and doripenem) and ceftazidime were increased by 8 to 27% in the presence of 32 microg/ml of ME1071. The corresponding resistance rates were decreased by 13 to 46%, respectively. On the other hand, ME1071 showed weaker or no potentiation with non-MBL producers. The K(i) value of ME1071 for IMP-1 was 0.4 microM, significantly lower than the K(m) values of carbapenems for the IMP-1 enzyme. On the other hand, the K(i) value of ME1071 for VIM-2 was 120 microM, higher than the K(m) values of carbapenems for the VIM-2 enzyme. Results of this study indicate that ME1071 can potentiate the activity of ceftazidime and carbapenems against MBL-producing strains of P. aeruginosa.
Sigma-Aldrich
Thioglycolic acid solution, ~70 % (w/w) in H2O
Supelco
Thioglycolic acid solution, ~80% in H2O, for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%
Sigma-Aldrich
Thioglycolic acid, ≥99%
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)
Sigma-Aldrich
Thioglycolic acid, 98%