MilliporeSigma
  • Analysis of the lipid extraction performance in a cascade process for Scenedesmus almeriensis biorefinery.
Analysis of the lipid extraction performance in a cascade process for Scenedesmus almeriensis biorefinery.
Biotechnology for biofuels (2021-01-16)
I Papachristou, S Akaberi, A Silve, E Navarro-López, R Wüstner, K Leber, N Nazarova, G Müller, W Frey
ABSTRACT
Microalgae have attracted considerable interest due to their ability to produce a wide range of valuable compounds. Pulsed Electric Fields (PEF) has been demonstrated to effectively disrupt the microalgae cells and facilitate intracellular extraction. To increase the commercial viability of microalgae, the entire biomass should be exploited with different products extracted and valorized according to the biorefinery scheme. However, demonstrations of multiple component extraction in series are very limited in literature. This study aimed to develop an effective lipid extraction protocol from wet Scenedesmus almeriensis after PEF-treatment with 1.5 MJ·kgDW-1. A cascade process, i.e., the valorization of several products in row, was tested with firstly the collection of the released carbohydrates in the water fraction, then protein enzymatic hydrolysis and finally lipid extraction. Biomass processed with high pressure homogenization (HPH) on parallel, served as benchmark. Lipid extraction with ethanol:hexane (1:0.41 vol/vol) offered the highest yields from the different protocols tested. PEF-treatment promoted extraction with almost 70% of total lipids extracted against 43% from untreated biomass. An incubation step after PEF-treatment, further improved the yields, up to 83% of total lipids. Increasing the solvent volume by factor 2 offered no improvement. In comparison, extraction with two other systems utilizing only ethanol at room temperature or elevated at 60 °C were ineffective with less than 30% of total lipids extracted. Regarding cascade extraction, carbohydrate release after PEF was detected albeit in low concentrations. PEF-treated samples displayed slightly better kinetics during the enzymatic protein hydrolysis compared to untreated or HPH-treated biomass. The yields from a subsequent lipid extraction were not affected after PEF but were significantly increased for untreated samples (66% of total lipids), while HPH displayed the lowest yields (~ 49% of total lipids). PEF-treatment successfully promoted lipid extraction from S. almeriensis but only in combination with a polar:neutral co-solvent (ethanol:hexane). After enzymatic protein hydrolysis in cascade processing; however, untreated biomass displayed equal lipid yields due to the disruptive effect of the proteolytic enzymes. Therefore, the positive impact of PEF in this scheme is limited on the improved reaction kinetics exhibited during the enzymatic hydrolysis step.