MilliporeSigma
  • A class of novel conjugates of substituted purine and Gly-AA-OBzl: synthesis and evaluation of orally analgesic activity.
A class of novel conjugates of substituted purine and Gly-AA-OBzl: synthesis and evaluation of orally analgesic activity.
Bioorganic & medicinal chemistry letters (2009-05-24)
Guifeng Kang, Ming Zhao, Xiaoyi Zhang, Li Peng, Chunbo Li, Wei Mao, Weidong Ye, Shiqi Peng
ABSTRACT
Aimed at the chemotherapy of chronic pain two kinds of analgesic pharmacophores, substituted purine and Gly-AA-OBzl, were coupled via a five-step-reaction procedure and 19 novel conjugates N-[2-chloro-9-(tetrahydropyran-2-yl)-9H-purin-6-yl]-N-cyclopropylglycylamino acid benzylesters were provided. On mouse-tail flick model their in vivo analgesic activities were assayed. The results indicate that introducing Gly-OC(2)H(5) into the 6-position of the substituted purine leads to ambiguous increase of the analgesic activity, while introducing Gly-AA-OBzl into this position leads to significant increase of the analgesic activity.
Sigma-Aldrich
2,6-Dichloropurine, 97%
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)