MilliporeSigma
  • A strategy for repression of arsenic toxicity through nuclear factor E2 related factor 2 activation mediated by the (E)-2-alkenals in Coriandrum sativum L. leaf extract.
A strategy for repression of arsenic toxicity through nuclear factor E2 related factor 2 activation mediated by the (E)-2-alkenals in Coriandrum sativum L. leaf extract.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-09-02)
Yumi Abiko, Miyuki Okada, Hanako Aoki, Mai Mizokawa, Yoshito Kumagai
ABSTRACT
Activation of the Kelch-like ECH-associated protein 1 (Keap1)/nuclear factor E2 related factor 2 (Nrf2) system plays a role in repression of xenobiotic toxicity. The Coriandrum sativum L. leaf extract (CSLE) contains various aliphatic electrophiles such as (E)-2-decenal and (E)-2-dodecenal. In the present study, we examined the activation of Nrf2 coupled to chemical modification of Keap1 mediated by (E)-2-alkenals in CSLE, and the protective role of CSLE and (E)-2-alkenals against inorganic arsenite (iAsIII) cytotoxicity. Ultra-performance liquid chromatography-elevated collision energy mass spectrometry analysis revealed that (E)-2-decenal modified recombinant Keap1 at Cys241, Cys249, Cys257 and His274. Exposure of HepG2 cells to CSLE, (E)-2-decenal, or (E)-2-dodecenal upregulated Nrf2-related downstream signaling such as expression of phase-II xenobiotic-metabolizing enzymes and phase-III transporters involved in cytoprotection against iAsIII. Pretreatment with CSLE or (E)-2-butenal, a prototype of (E)-2-alkenal, prior to iAsIII exposure suppressed accumulation of iAsIII significantly and reduced iAsIII-induced cytotoxicity in cells. Oral administration of CSLE to C57BL/6 mice upregulated downstream proteins of Nrf2 and reduced accumulation of arsenic in liver tissue. The present study indicates that CSLE containing (E)-2-alkenals activates Nrf2, leading to a reduction in arsenic accumulation in vivo.
Sigma-Aldrich
trans-2-Dodecenal, ≥93%, stabilized, FG
Pricing and availability is not currently available.