MilliporeSigma
  • Synthesis, stereochemical separation, and biological evaluation of selective inhibitors of human MAO-B: 1-(4-arylthiazol-2-yl)-2-(3-methylcyclohexylidene)hydrazines.
Synthesis, stereochemical separation, and biological evaluation of selective inhibitors of human MAO-B: 1-(4-arylthiazol-2-yl)-2-(3-methylcyclohexylidene)hydrazines.
Journal of medicinal chemistry (2010-08-19)
Franco Chimenti, Daniela Secci, Adriana Bolasco, Paola Chimenti, Arianna Granese, Simone Carradori, Matilde Yáñez, Francisco Orallo, M Luisa Sanna, Bruno Gallinella, Roberto Cirilli
ABSTRACT
Novel 1-(4-arylthiazol-2-yl)-2-(3-methylcyclohexylidene)hydrazine derivatives have been investigated for their ability to inhibit selectively the activity of the human B isoform of monoamine oxidase. These compounds were obtained as racemates and (R)-enantiomers by a stereoconservative synthetic pattern in high yield and enantiomeric excess. The (S)-enantiomers of the most active derivatives have been separated by enantioselective HPLC. All compounds showed selective activity against hMAO-B with IC(50) ranging between 21.90 and 0.018 microM.
Sigma-Aldrich
Isatin, 97%