MilliporeSigma
  • Hereditary spastic paraplegia SPG8 mutations impair CAV1-dependent, integrin-mediated cell adhesion.
Hereditary spastic paraplegia SPG8 mutations impair CAV1-dependent, integrin-mediated cell adhesion.
Science signaling (2020-01-09)
Seongju Lee, Hyungsun Park, Peng-Peng Zhu, Soon-Young Jung, Craig Blackstone, Jaerak Chang
ABSTRACT
Mutations in WASHC5 (also known as KIAA0196) cause autosomal dominant hereditary spastic paraplegia (HSP) type SPG8. WASHC5, commonly called strumpellin, is a core component of the Wiskott-Aldrich syndrome protein and SCAR homolog (WASH) complex that activates actin nucleation at endosomes. Although various other cellular roles for strumpellin have also been described, none account for how SPG8-associated mutations lead to HSP. Here, we identified protein interactors of the WASH complex by immunoprecipitation and mass spectrometry and assessed the functions of strumpellin in cultured cells using both overexpression and RNA interference along with cell-spreading assays to investigate cell adhesion. We uncovered a decrease in CAV1 protein abundance as well as endosomal fission defects resulting from pathogenic SPG8 mutations. CAV1, a key component of caveolae, interacted with strumpellin in cells, and strumpellin inhibited the lysosomal degradation of CAV1. SPG8-associated missense mutations in strumpellin did not rescue endosomal tubulation defects, reduction in CAV1 protein abundance, or integrin-mediated cell adhesion in strumpellin-deficient cells. Mechanistically, we demonstrated that the WASH complex maintained CAV1 and integrin protein amounts by inhibiting their lysosomal degradation through its endosomal actin nucleation activity. In addition, the interaction of strumpellin with CAV1 stimulated integrin recycling, thereby promoting cell adhesion. These findings provide a molecular link between WASHC5 mutations and impairment of CAV1- and integrin-mediated cell adhesion, providing insights into the cellular pathogenesis of SPG8.
Sigma-Aldrich
Bafilomycin A1
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture