MilliporeSigma
  • Visualization of live, mammalian neurons during Kainate-infusion using magnetic resonance microscopy.
Visualization of live, mammalian neurons during Kainate-infusion using magnetic resonance microscopy.
NeuroImage (2020-06-04)
Jeremy J Flint, Kannan Menon, Brian Hansen, John Forder, Stephen J Blackband
ABSTRACT
Since its first description and development in the late 20th century, diffusion magnetic resonance imaging (dMRI) has proven useful in describing the microstructural details of biological tissues. Signal generated from the protons of water molecules undergoing Brownian motion produces contrast based on the varied diffusivity of tissue types. Images employing diffusion contrast were first used to describe the diffusion characteristics of tissues, later used to describe the fiber orientations of white matter through tractography, and most recently proposed as a functional contrast method capable of delineating neuronal firing in the active brain. Thanks to the molecular origins of its signal source, diffusion contrast is inherently useful at describing features of the microenvironment; however, limitations in achievable resolution in magnetic resonance imaging (MRI) scans precluded direct visualization of tissue microstructure for decades following MRI's inception as an imaging modality. Even after advancements in MRI hardware had permitted the visualization of mammalian cells, these specialized systems could only accommodate fixed specimens that prohibited the observation and characterization of physiological processes. The goal of the current study was to visualize cellular structure and investigate the subcellular origins of the functional diffusion contrast mechanism (DfMRI) in living, mammalian tissue explants. Using a combination of ultra-high field spectrometers, micro radio frequency (RF) coils, and an MRI-compatible superfusion device, we are able to report the first live, mammalian cells-α-motor neurons-visualized with magnetic resonance microscopy (MRM). We are also able to report changes in the apparent diffusion of the stratum oriens within the hippocampus-a layer comprised primarily of pyramidal cell axons and basal dendrites-and the spinal cord's ventral horn following exposure to kainate.
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Kainic acid monohydrate, ≥99% (TLC)