MilliporeSigma
  • Wavelength- and irradiance-dependent changes in intracellular nitric oxide level.
Wavelength- and irradiance-dependent changes in intracellular nitric oxide level.
Journal of biomedical optics (2020-08-14)
Nathaniel J Pope, Samantha M Powell, Jeffrey C Wigle, Michael L Denton
ABSTRACT
Photobiomodulation (PBM) refers to the beneficial effects of low-energy light absorption. Although there is a large body of literature describing downstream physiological benefits of PBM, there is a limited understanding of the molecular mechanisms underlying these effects. At present, the most popular hypothesis is that light absorption induces release of nitric oxide (NO) from the active site of cytochrome c oxidase (COX), allowing it to bind O2 instead. This is believed to increase mitochondrial respiration, and result in greater overall health of the cell due to increased adenosine triphosphate production. Although NO itself is a powerful signaling molecule involved in a host of biological responses, less attention has been devoted to NO mechanisms in the context of PBM. The purpose of our work is to investigate wavelength-specific effects on intracellular NO release in living cells. We have conducted in-depth dosimetry analyses of NO production and function in an in vitro retinal model in response to low-energy exposure to one or more wavelengths of laser light. We found statistically significant wavelength-dependent elevations (10% to 30%) in intracellular NO levels following laser exposures at 447, 532, 635, or 808 nm. Sequential or simultaneous exposures to light at two different wavelengths enhanced the NO modulation up to 50% of unexposed controls. Additionally, the immediate increases in cellular NO levels were independent of the function of NO synthase, depended greatly on the substrate source of electrons entering the electron transport chain, and did not result in increased levels of cyclic guanosine monophosphate. Our study concludes the simple model of light-mediated release of NO from COX is unlikely to explain the wide variety of PBM effects reported in the literature. Our multiwavelength method provides a novel tool for studying immediate and early mechanisms of PBM as well as exploring intracellular NO signaling networks.
Sigma-Aldrich
SKF-525A, Hydrochloride, Cell permeable. Blocks glibenclamide-sensitive K+ channels. Inhibits neuronal nitric oxide synthase. Also inhibits hepatic drug metabolism by inhibiting the cytochrome P450 system.