MilliporeSigma
  • Cancer preventive agents 9. Betulinic acid derivatives as potent cancer chemopreventive agents.
Cancer preventive agents 9. Betulinic acid derivatives as potent cancer chemopreventive agents.
Bioorganic & medicinal chemistry letters (2009-06-02)
Kyoko Nakagawa-Goto, Koji Yamada, Masahiko Taniguchi, Harukuni Tokuda, Kuo-Hsiung Lee
ABSTRACT
C-3 esterifications of betulinic acid (BA, 1) and its A-ring homolog, ceanothic acid (CA, 2), were carried out to provide sixteen terpenoids, 4-19, including nine new compounds (4-12). All synthesized compounds were evaluated in an in vitro antitumor-promoting assay using the Epstein-Barr virus early antigen (EBV-EA) activation induced by 12-O-tetradecanoylphorbol-13-acetate (TPA) in Raji cells. Among them, compounds 4-6, 11-14, 16, and 17 displayed remarkable inhibitory effects of EBV-EA activation. BA analog 6, which contains a prenyl-like group, showed the most potent inhibitory effect (100%, 76%, 37%, and 11% inhibition of EBA activation at 1000, 500, 100, and 10mol ratio/TPA, respectively, with IC(50) value of 285mol ratio/32pmol TPA). Compound 6 merits further development as a cancer preventive agent.
Sigma-Aldrich
Betulinic acid, technical grade, 90%
Sigma-Aldrich
Betulinic acid, ≥98% (HPLC)