MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Design and synthesis of androgen receptor antagonists with bulky side chains for overcoming antiandrogen resistance.
Design and synthesis of androgen receptor antagonists with bulky side chains for overcoming antiandrogen resistance.
Journal of medicinal chemistry (2009-09-04)
Jinming Zhou, Guoyan Geng, Qingwen Shi, Francoise Sauriol, Jian Hui Wu
ABSTRACT
Incorporation of curcumin and beta-ionone into one chemical entity led to identification of a novel antiandrogen with two bulky side chains, 6, which is a pure antagonist of the wild-type and the T877A, W741C, and H874Y mutated androgen receptors (AR), showing no cross-reactivity with progesterone receptor and low micromolar cytotoxicity in LNCaP, PCa-2b, 22Rv1, and C4-2B prostate cancer cells. Molecular modeling indicates 6 adopts a "Y"-shape conformation and forms multiple hydrogen bonds with AR backbone.
Sigma-Aldrich
β-Ionone, predominantly trans, ≥97%, FCC, FG
Sigma-Aldrich
β-Ionone, 96%