MilliporeSigma
  • A Novel BCL-2 Inhibitor APG-2575 Exerts Synthetic Lethality With BTK or MDM2-p53 Inhibitor in Diffuse Large B-Cell Lymphoma.
A Novel BCL-2 Inhibitor APG-2575 Exerts Synthetic Lethality With BTK or MDM2-p53 Inhibitor in Diffuse Large B-Cell Lymphoma.
Oncology research (2020-02-26)
Qiuyun Luo, Wentao Pan, Suna Zhou, Guangfeng Wang, Hanjie Yi, Lin Zhang, Xianglei Yan, Luping Yuan, Zhenyi Liu, Jing Wang, Haibo Chen, MiaoZhen Qiu, DaJun Yang, Jian Sun
ABSTRACT
Despite therapeutic advances, the effective treatment for relapsed or refractory diffuse large B-cell lymphoma (DLBCL) remains a major clinical challenge. Evasion of apoptosis through upregulating antiapoptotic B-cell lymphoma-2 (BCL-2) family members and p53 inactivation, and abnormal activation of B-cell receptor signaling pathway are two important pathogenic factors for DLBCL. In this study, our aim is to explore a rational combination of BCL-2 inhibitor plus Brutons tyrosine kinase (BTK) blockade or p53 activation for treating DLBCL with the above characteristics. We demonstrated that a novel BCL-2 selective inhibitor APG-2575 effectively suppressed DLBCL with BCL-2 high expression via activating the mitochondrial apoptosis pathway. BTK inhibitor ibrutinib combined with BCL-2 inhibitors showed synergistic antitumor effect in DLBCL with mean expression of BCL-2 and myeloid cell leukemia-1 (MCL-1) through upregulating the expression level of BIM and modulating MCL-1 and p-Akt expression. For p53 wild-type DLBCL with high expression of BCL-2, APG-2575 showed strong synergic effect with mouse double minute 2 (MDM2)p53 inhibitor APG-115 that can achieve potent antitumor effect and markedly prolong survival in animal models. Collectively, our data provide an effective and precise therapeutic strategy through rational combination of BCL-2 and BTK or MDM2p53 inhibitors for DLBCL, which deserves further clinical investigation.