MilliporeSigma
  • Synthesis and structure-activity relationships of 8-(pyrid-3-yl)pyrazolo[1,5-a]-1,3,5-triazines: potent, orally bioavailable corticotropin releasing factor receptor-1 (CRF1) antagonists.
Synthesis and structure-activity relationships of 8-(pyrid-3-yl)pyrazolo[1,5-a]-1,3,5-triazines: potent, orally bioavailable corticotropin releasing factor receptor-1 (CRF1) antagonists.
Journal of medicinal chemistry (2009-04-14)
Paul J Gilligan, Todd Clarke, Liqi He, Snjezana Lelas, Yu-Wen Li, Karen Heman, Lawrence Fitzgerald, Keith Miller, Ge Zhang, Anne Marshall, Carol Krause, John F McElroy, Kathyrn Ward, Kim Zeller, Harvey Wong, Steven Bai, Joanne Saye, Scott Grossman, Robert Zaczek, Stephen P Arneric, Paul Hartig, David Robertson, George Trainor
ABSTRACT
This report describes the syntheses and structure-activity relationships of 8-(substituted pyridyl)pyrazolo[1,5-a]-1,3,5-triazine corticotropin releasing factor receptor-1 (CRF(1)) receptor antagonists. These CRF(1) receptor antagonists may be potential anxiolytic or antidepressant drugs. This research resulted in the discovery of compound 13-15, which is a potent, selective CRF(1) antagonist (hCRF(1) IC(50) = 6.1 +/- 0.6 nM) with weak affinity for the CRF-binding protein and biogenic amine receptors. This compound also has a good pharmacokinetic profile in dogs. Analogue 13-15 is orally effective in two rat models of anxiety: the defensive withdrawal (situational anxiety) model and the elevated plus maze test. Analogue 13-15 has been advanced to clinical trials.
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Furosemide