MilliporeSigma
  • Cobalt(II) Diphenylazodioxide Complexes Induce Apoptosis in SK-HEP-1 Cells.
Cobalt(II) Diphenylazodioxide Complexes Induce Apoptosis in SK-HEP-1 Cells.
ACS omega (2019-09-19)
Norah J Alghamdi, Lakshmi Balaraman, Kylin A Emhoff, Ahmed M H Salem, Ruhan Wei, Aimin Zhou, W Christopher Boyd
ABSTRACT
The cobalt(II) complex salts [Co(bpy)(az)2](PF6)2 and [Co(az)4](PF6), each bearing the unusual cis-N,N'-diphenylazodioxide ligand, were both screened as possible anticancer agents against SK-HEP-1 liver cancer cells. Both compounds were found to induce substantial apoptosis as an increasing function of concentration and time. Measurement of apoptosis-related proteins indicated that both the extrinsic and intrinsic pathways of apoptosis were activated. The apoptotic activity induced by these salts is not displayed either by simple cobalt(II) salts or complexes or by the free nitrosobenzene ligand. Additionally, these compounds did not induce apoptosis, as assessed by poly(adenosine diphosphate-ribose) polymerase cleavage, in several other cell lines.
Sigma-Aldrich
Fluorometric Intracellular Ros Kit, sufficient for 200 fluorometric tests (green)
Pricing and availability is not currently available.