MilliporeSigma
  • Sex-specific correlation of IGFBP-2 and IGFBP-3 with vitamin D status in adults with obesity: a cross-sectional serum proteomics study.
Sex-specific correlation of IGFBP-2 and IGFBP-3 with vitamin D status in adults with obesity: a cross-sectional serum proteomics study.
Nutrition & diabetes (2018-10-06)
Nasser M Al-Daghri, Antigoni Manousopoulou, Majed S Alokail, Sobhy Yakout, Amal Alenad, Diana J Garay-Baquero, Miltiadis Fotopoulos, Jie Teng, Omar Al-Attas, Yousef Al-Saleh, Shaun Sabico, George P Chrousos, Spiros D Garbis
ABSTRACT
Subjects with low vitamin D levels are at risk of cardiometabolic disease. The aim of this study was to identify novel serological markers linking vitamin D status with cardiometabolic profile in non-diabetic adults with obesity. For the discovery phase, we used quantitative serum proteomics in sex-matched, age-matched and BMI-matched subjects with obesity [BMI: 25-35 kg/m2] and low [25(OH)D < 50 nmol/L] vs. high vitamin D status [25(OH)D > 50 nmol/L] (n = 16). For the validation phase, we performed ELISA in a larger cohort with similar characteristics (n = 179). We identified 423 and 549 differentially expressed proteins in the high vs. low vitamin D groups of the male and female cohorts, respectively. The small molecule biochemistry protein networks and the glycolysis|gluconeogenesis pathway were significantly enriched in the DEPs of both sexes. As surrogate markers to these processes, the insulin-like growth factor binding protein -2  (IGFBP-2) was upregulated in males, whereas   IGFBP-3 was upregulated in females from the high Vitamin D status. This sex-specific trend  was confirmed using Luminex ELISA to an independent but clinically analogous cohort of males (n = 84, p = 0.002) and females (n = 95, p = 0.03). The high Vitamin D status correlated with the serological upregulation of IGFBP-2 in males and IGFBP-3 in females with obesity and may constitute surrogate markers of risk reduction of cardiometabolic disease.
Product Description
Millipore
MILLIPLEX® Human IGF Binding Protein Magnetic Bead Panel - Metabolism Multiplex Assay, The Human IGF Binding Protein Panel, using the Luminex xMAP technology, enables the simultaneous analysis of 7 IGFBP protein biomarkers in human serum, plasma and cell culture samples.