MilliporeSigma
  • Neolignans, a coumarinolignan, lignan derivatives, and a chromene: anti-inflammatory constituents from Zanthoxylum avicennae.
Neolignans, a coumarinolignan, lignan derivatives, and a chromene: anti-inflammatory constituents from Zanthoxylum avicennae.
Journal of natural products (2008-01-24)
Jih-Jung Chen, Tzu-Ying Wang, Tsong-Long Hwang
ABSTRACT
Eight new compounds, including four new neolignans, (7' S,8' S)-bilagrewin ( 1), (7' S,8' S)-5-demethoxybilagrewin ( 2), (7' S,8' S)-5- O-demethyl-4'- O-methylbilagrewin ( 3), and (7' S,8' S)-nocomtal ( 4), a new coumarinolignan, (7' S,8' S)-4'- O-methylcleomiscosin D ( 5), two new lignan derivatives, (+)-9'- O-( Z)-feruloyl-5,5'-dimethoxylariciresinol ( 6) and (+)-9'- O-( E)-feruloyl-5,5'-dimethoxylariciresinol ( 7), and a new chromene, ( E)-3-(2,2-dimethyl-2 H-chromen-6-yl)prop-2-enal ( 8), have been isolated from the stem wood of Zanthoxylum avicennae, together with 18 known compounds ( 9- 26). The structures of these new compounds were determined through spectroscopic and MS analyses. (7' S,8' S)-4'- O-Methylcleomiscosin D ( 5), cleomiscosin D ( 9), skimmianine ( 18), robustine ( 19), and integrifoliolin ( 23) exhibited inhibition (IC 50 < or = 18.19 microM) of superoxide anion generation by human neutrophils in response to formyl- l-methionyl- l-leucyl- l-phenylalanine/cytochalasin B (FMLP/CB). In addition, skimmianine ( 18) inhibited FMLP/CB-induced elastase release with an IC 50 value of 19.15 +/- 0.66 microM.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)
Sigma-Aldrich
Scopoletin, ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)