MilliporeSigma
  • VacA, cagA, iceA and oipA genotypes status and antimicrobial resistance properties of Helicobacter pylori isolated from various types of ready to eat foods.
VacA, cagA, iceA and oipA genotypes status and antimicrobial resistance properties of Helicobacter pylori isolated from various types of ready to eat foods.
Annals of clinical microbiology and antimicrobials (2016-01-23)
Behsan Hemmatinezhad, Hassan Momtaz, Ebrahim Rahimi
ABSTRACT
Despite the high clinical standing of Helicobacter pylori, its exact routes of transmission and origin have not been determined. Based on the contentious hypothesis, foods play an important roles in the transmission of H. pylori to humans. The present study was carried out to investigate the vacA, cagA, oipA and iceA genotypes status of H. pylori isolated from the various types of ready to eat foods. A total of 550 ready to eat food samples were cultured and tested. H. pylori-positive strains were analyzed for the presence of various genotypes and antimicrobial resistance pattern. Seventy four out of 550 (13.45 %) samples were positive for H. pylori. Olvie salad (36 %), restaurant salad (30 %), fruit salad (28 %) and soup (22 %) were the most commonly contaminated. H. pylori strains harbored the highest levels of resistance against amoxicillin (94.59 %), ampicillin (93.24 %), metronidazole (89.18 %) and tetracycline (72.97 %). The most commonly detected genotypes were vacA s1a (78.37 %), vacA m2 (75.67 %), vacA m1a (51.35 %) and cagA (41.89 %). The prevalence of iceA1, iceA2 and oipA genotypes were 13.51, 4.05 and 18.91 %, respectively. S1am2 (70.27 %), s1am1a (39.18 %) and m1am2 (31.08 %) were the most commonly detected combined genotypes. Of 40 different genotypic combinations, s1a/cagA+/iceA1/oipA- (12.16 %), s1a/cagA+/iceA1/oipA+ (10.81 %) and s1a/cagA-/iceA1/oipA+ (10.81 %) were the most prevalent. The present investigation showed that some types of ready to eat food samples maybe the sources of resistant and virulent strains of H. pylori. Warily use of antibiotics with respect to the results of disk diffusion method and careful health monitoring on food and staffs of food producing companies maybe reduce the risk of H. pylori in foods.
Product Description
Roche
DNA Isolation Kit for Cells and Tissues, sufficient for 10 isolation(s)