MilliporeSigma
  • Design and microwave-assisted synthesis of 5-trifluoromethyl-4,5-dihydro-1H-pyrazoles: novel agents with analgesic and anti-inflammatory properties.
Design and microwave-assisted synthesis of 5-trifluoromethyl-4,5-dihydro-1H-pyrazoles: novel agents with analgesic and anti-inflammatory properties.
European journal of medicinal chemistry (2007-09-25)
Patricia D Sauzem, Pablo Machado, Maribel A Rubin, Gabriela da S Sant'anna, Henrique B Faber, Alessandra H de Souza, Carlos F Mello, Paulo Beck, Robert A Burrow, Helio G Bonacorso, Nilo Zanatta, Marcos A P Martins
ABSTRACT
In this work, we reported the synthesis and evaluation of the analgesic and anti-inflammatory properties of novel 3- or 4-substituted 5-trifluoromethyl-5-hydroxy-4,5-dihydro-1H-1-carboxyamidepyrazoles (where 3-/4-substituent=H/H, Me/H, Et/H, Pr/H, i-Pr/H, Bu/H, t-Bu/H, Ph/H, 4-Br-Ph/H and H/Me) designed in the exploration of the bioisosteric replacement of benzene present in salicylamide with a 5-trifluoromethyl-4,5-dihydro-1H-pyrazole scaffold. Target compounds were synthesized from the cyclocondensation of 4-alkoxy-1,1,1-trifluoromethyl-3-alken-2-ones with semicarbazide hydrochloride through a rapid one-pot reaction via microwave irradiation. In addition to spectroscopic data, the structure of the compounds was supported by X-ray diffraction. Subcutaneous administration of the 5-trifluoromethyl-4,5-dihydro-1H-pyrazoles decreased pain-related behavior during neurogenic and inflammatory phases of the formalin test in mice. Moreover, the more active analgesic compounds (3-/4-=Et/H and H/Me) significantly decreased carrageenan-induced paw edema in mice. The data obtained in this work suggest that the synthesized compounds could be promising candidates for the future development of novel analgesic and anti-inflammatory agents.
Sigma-Aldrich
Salicylamide, 99%