MilliporeSigma
  • Axial chirality and affinity at the GABA(A) receptor of pyrimido[1,2-a][1,4]benzodiazepines and related compounds.
Axial chirality and affinity at the GABA(A) receptor of pyrimido[1,2-a][1,4]benzodiazepines and related compounds.
Bioorganic & medicinal chemistry (2008-10-03)
Shoukou Lee, Tomonori Kamide, Hidetsugu Tabata, Hideyo Takahashi, Motoo Shiro, Hideaki Natsugari
ABSTRACT
The pyrimido[1,2-a][1,4]benzodiazepines (1a-c) and the 8-membered analogues (diazocines 2a and 2b) were separated into their atropisomers with HPLC on a chiral column. High stereochemical stability was observed in the atropisomer of the 8-membered derivatives (2a and 2b), and the 1,4-benzodiazepine (1c) with 2'-chloro at the pendant phenyl showed a lower energy barrier for the conversion between the atropisomers compared with that with the unsubstituted pendant phenyl (1a). The aR isomer of 1a-c was revealed to be the eutomer in GABA(A) receptor binding, and the eutomer 1c-R showed extremely potent activity with an IC(50) value of 1.5 nM.
Sigma-Aldrich
Diazepam