MilliporeSigma
  • Enhancement of the anti-tumour effect of cyclophosphamide by the bioreductive drugs AQ4N and tirapazamine.
Enhancement of the anti-tumour effect of cyclophosphamide by the bioreductive drugs AQ4N and tirapazamine.
British journal of cancer (2000-04-26)
O P Friery, R Gallagher, M M Murray, C M Hughes, E S Galligan, I A McIntyre, L H Patterson, D G Hirst, S R McKeown
ABSTRACT
The ability of the bioreductive drugs AQ4N and tirapazamine to enhance the anti-tumour effect of cyclophosphamide was assessed in three murine tumour models. In male BDF mice implanted with the T50/80 mammary carcinoma, AQ4N (50-150 mg kg(-1)) in combination with cyclophosphamide (100 mg kg(-1)) produced an effect equivalent to a single 200 mg kg 1 dose of cyclophosphamide. Tirapazamine (25 mg kg(-1)) in combination with cyclophosphamide (100 mg kg(-1)) produced an effect equivalent to a single 150 mg kg(-1) dose of cyclophosphamide. In C3H mice implanted with the SCCVII or RIF-1 tumours, enhancement of tumour cell killing was found with both drugs in combination with cyclophosphamide (50-200 mg kg(-1)); AQ4N (50-200 mg kg(-1)) produced a more effective combination than tirapazamine (12.5-50 mg kg(-1)). Unlike tirapazamine, which showed a significant increase in toxicity to bone marrow cells, the combination of AQ4N (100 mg kg(-1)) 6 h prior to cyclophosphamide (100 mg k(-1)) resulted in no additional toxicity towards bone marrow cells compared to that caused by cyclophosphamide alone. In conclusion, AQ4N gave a superior anti-tumour effect compared to tirapazamine when administered with a single dose of cyclophosphamide (100 mg kg(-1)).
Sigma-Aldrich
Banoxantrone dihydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.