MilliporeSigma
  • Genome-Wide Association Study of Event-Free Survival in Diffuse Large B-Cell Lymphoma Treated With Immunochemotherapy.
Genome-Wide Association Study of Event-Free Survival in Diffuse Large B-Cell Lymphoma Treated With Immunochemotherapy.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2015-10-16)
Hervé Ghesquieres, Susan L Slager, Fabrice Jardin, Amelie S Veron, Yan W Asmann, Matthew J Maurer, Thierry Fest, Thomas M Habermann, Marie C Bene, Anne J Novak, Sylvain Mareschal, Corinne Haioun, Thierry Lamy, Stephen M Ansell, Herve Tilly, Thomas E Witzig, George J Weiner, Andrew L Feldman, Ahmet Dogan, Julie M Cunningham, Curtis L Olswold, Thierry Jo Molina, Brian K Link, Noel Milpied, David G Cox, Gilles A Salles, James R Cerhan
ABSTRACT
We performed a multistage genome-wide association study to identify inherited genetic variants that predict outcome in diffuse large B-cell lymphoma patients treated with immunochemotherapy. We conducted a meta-analysis of two genome-wide association study data sets, one from the LNH2003B trial (N = 540), a prospective clinical trial from the Lymphoma Study Association, and the other from the Molecular Epidemiology Resource study (N = 312), a prospective observational study from the University of Iowa-Mayo Clinic Lymphoma Specialized Program of Research Excellence. Top single nucleotide polymorphisms were then genotyped in independent cohorts of patients from the Specialized Program of Research Excellence (N = 391) and the Groupe Ouest-Est des Leucémies Aiguës et Maladies du Sang (GOELAMS) -075 randomized trial (N = 294). We calculated the hazard ratios (HRs) and 95% CIs for event-free survival (EFS) and overall survival (OS) using a log-additive genetic model with adjustment for age, sex, and age-adjusted International Prognostic Index. In a meta-analysis of the four studies, the top loci for EFS were marked by rs7712513 at 5q23.2 (near SNX2 and SNCAIP; HR, 1.39; 95% CI, 1.23 to 1.57; P = 2.08 × 10(-7)), and rs7765004 at 6q21 (near MARCKS and HDAC2; HR, 1.38; 95% CI, 1.22 to 1.57; P = 7.09 × 10(-7)), although they did not reach conventional genome-wide significance (P = 5 × 10(-8)). Both rs7712513 (HR, 1.49; 95% CI, 1.29 to 1.72; P = 3.53 × 10(-8)) and rs7765004 (HR, 1.47; 95% CI, 1.27 to 1.71; P = 5.36 × 10(-7)) were also associated with OS. In exploratory analyses, a two-single nucleotide polymorphism risk score was highly predictive of EFS (P = 1.78 × 10(-12)) and was independent of treatment, IPI, and cell-of-origin classification. Our study provides encouraging evidence for associations between loci at 5q23.2 and 6q21 with EFS and OS in patients with diffuse large B-cell lymphoma treated with immunochemotherapy, suggesting novel biology and the potential contribution of host genetics to the prognosis of this aggressive malignancy.