MilliporeSigma
  • Kinetic analysis of the lactate-dehydrogenase-coupled reaction process and measurement of alanine transaminase by an integration strategy.
Kinetic analysis of the lactate-dehydrogenase-coupled reaction process and measurement of alanine transaminase by an integration strategy.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2010-11-17)
Xiaolan Yang, Beizhong Liu, Yu Sang, Yonghua Yuan, Jun Pu, Yin Liu, Zhirong Li, Juan Feng, Yanling Xie, Renkuan Tang, Huidong Yuan, Fei Liao
ABSTRACT
Kinetic analyses of lactate-dehydrogenase (LD)-coupled alanine transaminase (ALT) reaction processes were investigated for measuring ALT by an integration strategy. For measuring ALT by a kinetic analysis of an LD-coupled ALT reaction curve, candidate reaction curves were calculated via iterative numerical integration of the differential velocity equations to execute a weighted nonlinear-least-square-fitting. To realize the integration strategy, the conventional initial-velocity method was used if the ALT activities were below 25 U/L; otherwise, kinetic analyses of the reaction curves were employed. Of the reaction curves recorded at 10-s intervals, kinetic analyses gave ALT activities resistant to deviations in the LD kinetic parameters. The integration strategy yielded a higher value of the lower limit, but an upper limit of over 100 U/L by simulations and over 75 U/L with purified ALT. Also, its intra-run relative standard deviations were below 9% for 0.50 U/L ALT and below 5% for final 1 to 65 U/L ALT. The integration strategy gave consistent ALT activities in clinical sera. Hence, this new approach for kinetic analyses of ALT reaction processes and the integration strategy were effective to measure ALT.
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein