MilliporeSigma
  • Curcumin Derivative C086 Combined with Cisplatin Inhibits Proliferation of Osteosarcoma Cells.
Curcumin Derivative C086 Combined with Cisplatin Inhibits Proliferation of Osteosarcoma Cells.
Medical science monitor : international medical journal of experimental and clinical research (2020-08-01)
Xi Jiang, Yulin Huang
ABSTRACT
BACKGROUND Curcumin derivative C086 (cur C086) is a potential chemotherapeutic agent for patients with osteosarcoma. In this study, the effects of cur C086 combined with cisplatin on the biological processes of osteosarcoma cells were investigated. MATERIAL AND METHODS In this study, expression of BMIL1 was detected by real-time quantitative reverse transcription polymerase chain reaction and Western blotting in MG-63 cells treated with cur C086+cisplatin. Functions of cur C086+cisplatin on proliferation ability, apoptosis response, and metastatic potential of MG-63 cells were determined by MTT, flow cytometry, Hoechst 33258 staining and Transwell assays, respectively. In additionally, expression of P16, E-cadherin, epidermal growth factor (EGFR), and Notch1 was measured by Western blotting. RESULTS Expression of BMIL1 decreased significantly in MG-63 cells treated with cur C086 (20 µM)+cisplatin (1.28 nM). Treatment with cur C086+cisplatin considerably inhibited growth, migration, and invasion potential in MG-63 cells, whereas apoptosis was obviously upregulated. Moreover, cur C086+cisplatin suppressed BMIL1 expression or its potential downstream targets, P16, E-cadherin, EGFR, and Notch1. CONCLUSIONS The current results demonstrate that combined treatment with cur C086+cisplatin may be an effective form of chemotherapy for patients with osteosarcoma.
Sigma-Aldrich
MTT Formazan, MTT reduction end product
Pricing and availability is not currently available.