MilliporeSigma
  • Transition-Metal-Free Sulfuration/Annulation of Alkenes: Economical Access to Thiophenes Enabled by the Cleavage of Multiple C-H Bonds.
Transition-Metal-Free Sulfuration/Annulation of Alkenes: Economical Access to Thiophenes Enabled by the Cleavage of Multiple C-H Bonds.
Organic letters (2018-11-22)
Liang Chen, Hao Min, Weilan Zeng, Xiaoming Zhu, Yun Liang, Guobo Deng, Yuan Yang
ABSTRACT
A novel, atom economical, and transition-metal-free strategy for the synthesis of thiophenes from substituted buta-1-enes with potassium sulfide has been presented. The reaction achieves double C-S bond formations via cleavage of multiple C-H bonds and provides an efficient approach to access various functionalized thiophenes. Moreover, the strategy can also be used for the synthesis of thiophenes from 1,4-diaryl-1,3-dienes. Mechanistically, DMSO plays a role of oxidant and S3•- in situ generated from K2S is involved.
Sigma-Aldrich
trans,trans-1,4-Diphenyl-1,3-butadiene, 98%