MilliporeSigma
  • Leukadherin-1 ameliorates endothelial barrier damage mediated by neutrophils from critically ill patients.
Leukadherin-1 ameliorates endothelial barrier damage mediated by neutrophils from critically ill patients.
Journal of intensive care (2018-03-24)
Catherine M Dickinson, Brian W LeBlanc, Muhammad M Edhi, Daithi S Heffernan, Mohd Hafeez Faridi, Vineet Gupta, William G Cioffi, Xian O'Brien, Jonathan S Reichner
ABSTRACT
Multi-organ failure occurs during critical illness and is mediated in part by destructive neutrophil-to-endothelial interactions. The β2 integrin receptor, CR3 (complement receptor 3; Mac-1; CD11b/CD18), which binds endothelial intercellular adhesion molecule-1 (ICAM-1), plays a key role in promoting the adhesion of activated neutrophils to inflamed endothelia which, when prolonged and excessive, can cause vascular damage. Leukadherin-1 (LA-1) is a small molecule allosteric activator of CR3 and has been shown to promote adhesion of blood neutrophils to inflamed endothelium and restrict tissue infiltration. Therefore, LA-1 offers a novel mechanism of anti-inflammatory action by activation, rather than inhibition, of the neutrophil CR3 integrin. However, whether promotion of neutrophil-to-endothelial interaction by this novel therapeutic is of benefit or detriment to endothelial barrier function is not known. Critically ill septic and trauma patients were prospectively enrolled from the surgical and the trauma ICU. Blood was collected from these patients and healthy volunteers. Neutrophils were isolated by dextran sedimentation and adhered to TNF-α (tumor necrosis factor-α)-activated human umbilical vein endothelial (HUVEC) monolayers in the presence or absence of fMLP (formylmethionine-leucine-phenylalanine) and/or LA-1. Electric cell-substrate impedance sensing (ECIS) and exposure of underlying collagen were used to quantify endothelial barrier function and permeability. Neutrophils from critically ill trauma and septic patients caused similar degrees of endothelial barrier disruption which exceeded that caused by cells obtained from healthy controls both kinetically and quantitatively. LA-1 protected barrier function in the absence and presence of fMLP which served as a secondary stimulant to cause maximal loss of barrier function. LA-1 protection was also observed by quantifying collagen exposure underlying endothelial cells challenged with fMLP-stimulated neutrophils. LA-1 treatment resulted in decreased migration dynamics of neutrophils crawling on an endothelial monolayer with reduced speed (μm/s = 0.25 ± 0.01 vs. 0.06 ± 0.01, p < 0.05), path length (μm = 199.5 ± 14.3 vs. 42.1 ± 13.0, p < 0.05), and displacement (μm = 65.2 ± 4.7 vs. 10.4 ± 1.3; p < 0.05). Neutrophils from patients with trauma or sepsis cause endothelial barrier disruption to a similar extent relative to each other. The CR3 agonist LA-1 protects endothelial barrier function from damage caused by neutrophils obtained from both populations of critically ill patients even when exposed to secondary stimulation.
Sigma-Aldrich
Leukadherin-1, ≥98% (HPLC)