MilliporeSigma
  • Inter-familial and intra-familial phenotypic variability in three Sicilian families with Anderson-Fabry disease.
Inter-familial and intra-familial phenotypic variability in three Sicilian families with Anderson-Fabry disease.
Oncotarget (2017-10-06)
Antonino Tuttolomondo, Irene Simonetta, Giovanni Duro, Rosaria Pecoraro, Salvatore Miceli, Paolo Colomba, Carmela Zizzo, Antonia Nucera, Mario Daidone, Tiziana Di Chiara, Rosario Scaglione, Vittoriano Della Corte, Francesca Corpora, Danai Vogiatzis, Antonio Pinto
ABSTRACT
Anderson-Fabry disease (AFD) is an inborn lysosomal enzymopathy resulting from the deficient or absent activity of the lysosomal exogalactohydrolase, α-galactosidase A. This deficiency, results in the altered metabolism of glycosphingolipids which leads to their accumulation in lysosomes, thus to cellular and vascular dysfunction. To date, numerous mutations (according to recent data more than 1000 mutations) have been reported in the GLA intronic and exonic mutations. Traditionally, clinical manifestations are more severe in affected hemizygous males than in females. Nevertheless, recent studies have described severe organ dysfunction in women. This study reports clinical, biochemical, and molecular findings of the members of three Sicilian families. The clinical history of these patients highlights a remarkable interfamilial and intrafamilial phenotypic variability which characterizes Fabry disease relative to target organs and severity of clinical manifestations. Our findings, in agreement with previous data, report a little genotype-phenotype correlation for the disease, suggesting that the wide phenotypic variability of Anderson-Fabry disease is not completely ascribable to different gene mutations but other factors and mechanisms seem to be involved in the pathogenesis and clinical expression of the disease. Moreover, this study emphasies the importance of pedigree analysis in the family of each proband for identifying other possibly affected relatives.
Sigma-Aldrich
Deoxygalactonojirimycin hydrochloride
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphate-Citrate Buffer, tablet
Pricing and availability is not currently available.