MilliporeSigma
  • Discovery of potent SOS1 inhibitors that block RAS activation via disruption of the RAS-SOS1 interaction.
Discovery of potent SOS1 inhibitors that block RAS activation via disruption of the RAS-SOS1 interaction.
Proceedings of the National Academy of Sciences of the United States of America (2019-01-27)
Roman C Hillig, Brice Sautier, Jens Schroeder, Dieter Moosmayer, André Hilpmann, Christian M Stegmann, Nicolas D Werbeck, Hans Briem, Ulf Boemer, Joerg Weiske, Volker Badock, Julia Mastouri, Kirstin Petersen, Gerhard Siemeister, Jan D Kahmann, Dennis Wegener, Niels Böhnke, Knut Eis, Keith Graham, Lars Wortmann, Franz von Nussbaum, Benjamin Bader
ABSTRACT
Since the late 1980s, mutations in the RAS genes have been recognized as major oncogenes with a high occurrence rate in human cancers. Such mutations reduce the ability of the small GTPase RAS to hydrolyze GTP, keeping this molecular switch in a constitutively active GTP-bound form that drives, unchecked, oncogenic downstream signaling. One strategy to reduce the levels of active RAS is to target guanine nucleotide exchange factors, which allow RAS to cycle from the inactive GDP-bound state to the active GTP-bound form. Here, we describe the identification of potent and cell-active small-molecule inhibitors which efficiently disrupt the interaction between KRAS and its exchange factor SOS1, a mode of action confirmed by a series of biophysical techniques. The binding sites, mode of action, and selectivity were elucidated using crystal structures of KRASG12C-SOS1, SOS1, and SOS2. By preventing formation of the KRAS-SOS1 complex, these inhibitors block reloading of KRAS with GTP, leading to antiproliferative activity. The final compound 23 (BAY-293) selectively inhibits the KRAS-SOS1 interaction with an IC50 of 21 nM and is a valuable chemical probe for future investigations.
Sigma-Aldrich
BAY-293, ≥98% (HPLC)
Pricing and availability is not currently available.