MilliporeSigma
  • TIMP-2 mediated inhibition of angiogenesis: an MMP-independent mechanism.
TIMP-2 mediated inhibition of angiogenesis: an MMP-independent mechanism.
Cell (2003-07-31)
Dong-Wan Seo, Hongmei Li, Liliana Guedez, Paul T Wingfield, Tere Diaz, Rita Salloum, Bei-yang Wei, William G Stetler-Stevenson
ABSTRACT
Tissue inhibitors of metalloproteinases (TIMPs) suppress matrix metalloproteinase (MMP) activity critical for extracellular matrix turnover associated with both physiologic and pathologic tissue remodeling. We demonstrate here that TIMP-2 abrogates angiogenic factor-induced endothelial cell proliferation in vitro and angiogenesis in vivo independent of MMP inhibition. These effects require alpha 3 beta 1 integrin-mediated binding of TIMP-2 to endothelial cells. Further, TIMP-2 induces a decrease in total protein tyrosine phosphatase (PTP) activity associated with beta1 integrin subunits as well as dissociation of the phosphatase SHP-1 from beta1. TIMP-2 treatment also results in a concomitant increase in PTP activity associated with tyrosine kinase receptors FGFR-1 and KDR. Our findings establish an unexpected, MMP-independent mechanism for TIMP-2 inhibition of endothelial cell proliferation in vitro and reveal an important component of the antiangiogenic effect of TIMP2 in vivo.
Sigma-Aldrich
Rat TIMP-2 ELISA Kit, for cell culture supernatants, plasma, and serum samples