MilliporeSigma
  • Mitochondrial phosphate-carrier deficiency: a novel disorder of oxidative phosphorylation.
Mitochondrial phosphate-carrier deficiency: a novel disorder of oxidative phosphorylation.
American journal of human genetics (2007-02-03)
Johannes A Mayr, Olaf Merkel, Sepp D Kohlwein, Boris R Gebhardt, Hansjosef Böhles, Ulrike Fötschl, Johannes Koch, Michaela Jaksch, Hanns Lochmüller, Rita Horváth, Peter Freisinger, Wolfgang Sperl
ABSTRACT
The mitochondrial phosphate carrier SLC25A3 transports inorganic phosphate into the mitochondrial matrix, which is essential for the aerobic synthesis of adenosine triphosphate (ATP). We identified a homozygous mutation--c.215G-->A (p.Gly72Glu)--in the alternatively spliced exon 3A of this enzyme in two siblings with lactic acidosis, hypertrophic cardiomyopathy, and muscular hypotonia who died within the 1st year of life. Functional investigation of intact mitochondria showed a deficiency of ATP synthesis in muscle but not in fibroblasts, which correlated with the tissue-specific expression of exon 3A in muscle versus exon 3B in fibroblasts. The enzyme defect was confirmed by complementation analysis in yeast. This is the first report of patients with mitochondrial phosphate-carrier deficiency.