MilliporeSigma
  • Continuous Online Protein Quality Monitoring during Perfusion Culture Production Using an Integrated Micro/Nanofluidic System.
Continuous Online Protein Quality Monitoring during Perfusion Culture Production Using an Integrated Micro/Nanofluidic System.
Analytical chemistry (2020-03-14)
Taehong Kwon, Sung Hee Ko, Jean-François P Hamel, Jongyoon Han
ABSTRACT
We demonstrate a new micro/nanofluidic system for continuous and automatic monitoring of protein product size and quantity directly from the culture supernatant during a high-cell-concentration CHO cell perfusion culture. A microfluidic device enables clog-free cell retention for a bench-scale (350 mL) perfusion bioreactor that continuously produces the culture supernatant containing monoclonal antibodies (IgG1). A nanofluidic device directly monitors the protein size and quantity in the culture supernatant. The continuous-flow and fully automated operation of this nanofluidic protein analytics reduces design complexity and offers more detailed information on protein products than offline and batch-mode conventional analytics. Moreover, chemical and mechanical robustness of the nanofluidic device enables continuous monitoring for several days to a week. This continuous and online protein quality monitoring could be deployed at different steps and scales of biomanufacturing to improve product quality and manufacturing efficiency.
Sigma-Aldrich
IgG1, Kappa from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)