MilliporeSigma
  • Mortality, oxidative stress and tau accumulation during ageing in parkin null mice.
Mortality, oxidative stress and tau accumulation during ageing in parkin null mice.
Journal of neurochemistry (2007-07-12)
Jose A Rodríguez-Navarro, M José Casarejos, Jaime Menéndez, Rosa M Solano, Izaskun Rodal, Ana Gómez, Justo García de Yébenes, Maria A Mena
ABSTRACT
Young parkin null (pk-/-) mice have subtle abnormalities of behaviour, dopamine (DA) neurotransmission and free radical production, but no massive loss of DA neurons. We investigated whether these findings are maintained while ageing. Pk-/- mice have reduced life span and age-related reduced exploratory behaviour, abnormal walking and posture, and behaviours similar to those of early Parkinson's disease (PD), reduced number of nigrostriatal DA neurons and proapoptotic shifts in the survival/death proteins in midbrain and striatum. Contrary to young pk-/- animals 24-month-old pk-/- mice do not have compensatory elevation of GSH in striatum, glutathione reductase (GR) and glutathione peroxidase (GPx) activities are increased and catalase unchanged. Aged pk-/- mice accumulate high levels of tau and fail to up-regulate CHIP and HSP70. Our results suggest that aged pk-/- mice lack of the compensatory mechanisms that maintain a relatively normal DA function in early adulthood. This study could help to explain the effects of ageing in patients with genetic risks for Parkinson's disease.
Sigma-Aldrich
Anti-Tau Antibody, a.a. 210-241, clone Tau-5, ascites fluid, clone Tau-5, Chemicon®