MilliporeSigma
  • Transcription factor Foxp1 regulates Foxp3 chromatin binding and coordinates regulatory T cell function.
Transcription factor Foxp1 regulates Foxp3 chromatin binding and coordinates regulatory T cell function.
Nature immunology (2019-01-16)
Catherine Konopacki, Yuri Pritykin, Yury Rubtsov, Christina S Leslie, Alexander Y Rudensky
ABSTRACT
Regulatory T cells (Treg cells), whose differentiation and function are controlled by transcription factor Foxp3, express the closely related family member Foxp1. Here we explored Foxp1 function in Treg cells. We found that a large number of Foxp3-bound genomic sites in Treg cells were occupied by Foxp1 in both Treg cells and conventional T cells (Tconv cells). In Treg cells, Foxp1 markedly increased Foxp3 binding to these sites. Foxp1 deficiency in Treg cells resulted in their impaired function and competitive fitness, associated with markedly reduced CD25 expression and interleukin-2 (IL-2) responsiveness, diminished CTLA-4 expression and increased SATB1 expression. The characteristic expression patterns of CD25, Foxp3 and CTLA-4 in Treg cells were fully or partially rescued by strong IL-2 signaling. Our studies suggest that Foxp1 serves an essential non-redundant function in Treg cells by enforcing Foxp3-mediated regulation of gene expression and enabling efficient IL-2 signaling in these cells.
Sigma-Aldrich
Anti-FoxP1 Antibody, serum, from rabbit
Pricing and availability is not currently available.