MilliporeSigma
Cytotoxic flavaglines and bisamides from Aglaia edulis.
Journal of natural products (2006-12-28)
Soyoung Kim, Young-Won Chin, Bao-Ning Su, Soedarsono Riswan, Leonardus B S Kardono, Johar J Afriastini, Heebyung Chai, Norman R Farnsworth, Geoffrey A Cordell, Steven M Swanson, A Douglas Kinghorn
ABSTRACT
Two new cyclopenta[b]benzofurans, aglaroxin A 1-O-acetate (2) and 3'-methoxyaglaroxin A 1-O-acetate (3), a new benzo[b]oxepine, 19,20-dehydroedulisone A (4), and five new cyclopenta[bc]benzopyrans, edulirin A (5), edulirin A 10-O-acetate (6), 19,20-dehydroedulirin A (7), isoedulirin A (8), and edulirin B (9), were isolated from the bark of Aglaia edulis, along with one known cyclopenta[b]benzofuran, aglaroxin A (1). Additionally, four new amides, aglamides A-D (10-13), as well as three known compounds, aglalactone, scopoletin, and 5-hydroxy-3,6,7,4'-tetramethoxyflavone, were isolated from the leaves and/or twigs of this species. The structures of the new compounds (2-13) were elucidated by interpretation of their spectroscopic data. All isolates obtained in this study were evaluated for cytotoxicity against both several human cancer cell lines (Lu1, LNCaP, and MCF-7) and a nontumorigenic (HUVEC) cell line. Among these isolates, the cyclopenta[b]benzofurans (1-3) exhibited potent in vitro cytotoxic activity (ED50 range 0.001 to 0.8 microg/mL). Aglaroxin A 1-O-acetate (2) was further evaluated in the in vivo P388 lymphocytic leukemia model, by intraperitoneal injection, but found to be inactive in this model.
Sigma-Aldrich
Scopoletin, ≥99%