MilliporeSigma
  • Carbonic anhydrase inhibitors. Novel sulfanilamide/acetazolamide derivatives obtained by the tail approach and their interaction with the cytosolic isozymes I and II, and the tumor-associated isozyme IX.
Carbonic anhydrase inhibitors. Novel sulfanilamide/acetazolamide derivatives obtained by the tail approach and their interaction with the cytosolic isozymes I and II, and the tumor-associated isozyme IX.
Bioorganic & medicinal chemistry letters (2004-12-18)
Hasan Turkmen, Mustafa Durgun, Serpil Yilmaztekin, Mahmut Emul, Alessio Innocenti, Daniela Vullo, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
A series of sulfonamides has been obtained by reacting sulfanilamide or 5-amino-1,3,4-thiadiazole-2-sulfonamide with omega-chloroalkanoyl chlorides, followed by replacement of the omega-chlorine atom with secondary amines. Tails incorporating heterocyclic amines belonging to the morpholine, piperidine and piperazine ring systems have been attached to these sulfonamides, by means of an alkanoyl-carboxamido linker containing from two to five carbon atoms. The new derivatives prepared in this way were tested as inhibitors of three carbonic anhydrase (CA, EC 4.2.1.1) isozymes, the cytosolic isozymes CA I and II, and the catalytic domain of the transmembrane, tumor-associated isozyme CA IX. Several low nanomolar CA I and CA II inhibitors were detected both in the aromatic and heterocyclic sulfonamide series, whereas the best hCA IX inhibitors (inhibition constants in the range of 22-35 nM) all belonged to the acetazolamide-like derivatives.
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, ≥98%