- 6-Hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid mimics active conformation of tyrosine in opioid peptides.
6-Hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid mimics active conformation of tyrosine in opioid peptides.
Bioorganic & medicinal chemistry letters (2005-05-03)
Emanuela Sperlinga, Piotr Kosson, Zofia Urbanczyk-Lipkowska, Giuseppe Ronsisvalle, Daniel B Carr, Andrzej W Lipkowski
PMID15863298
ABSTRACT
6-Hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid (6Htc) has been proposed as a rigid mimic of tyrosine conformation in opioid ligand-receptor complex. The significant receptor binding to mu and delta opioid receptors of respective analogues of deltorphin, dermorphin, and endomorphin with D,L-6Htc prove initial prediction.