MilliporeSigma
  • Synthesis and radioligand binding studies of C-5- and C-8-substituted 1-(3,4-dimethoxybenzyl)-2,2-dimethyl-1,2,3,4-tetrahydroisoquinoliniums as SK channel blockers related to N-methyl-laudanosine and N-methyl-noscapine.
Synthesis and radioligand binding studies of C-5- and C-8-substituted 1-(3,4-dimethoxybenzyl)-2,2-dimethyl-1,2,3,4-tetrahydroisoquinoliniums as SK channel blockers related to N-methyl-laudanosine and N-methyl-noscapine.
Journal of medicinal chemistry (2005-07-22)
Amaury Graulich, Jacqueline Scuvée-Moreau, Vincent Seutin, Jean-François Liégeois
ABSTRACT
The synthesis and the (125)I-apamin binding studies of original C-5- and C-8-substituted 1-(3,4-dimethoxy-benzyl)-2,2-dimethyl-1,2,3,4-tetrahydroisoquinoliniums and 1-(3,4-dimethoxy-benzyl)-6,6-dimethyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridiniums were performed in order to find a reversible and selective SK channel blocker structurally related to N-methyl-laudanosine and N-methyl-noscapine. A bulky alkyl substituent in the C-8 position of the tetrahydroisoquinoline produces a clear increase in the affinity for the apamin sensitive binding sites. The presence of an electron-withdrawing group in the C-5 and C-8 positions is not a suitable substitution for the affinity of drugs structurally related to N-methyl-laudanosine. Thiophenic analogues and 8-methoxy derivatives possess a poor affinity for the apamin sensitive binding sites. Electrophysiological studies performed with the most effective compound showed a blockade of the apamin sensitive afterhyperpolarization in rat dopaminergic neurons.
Sigma-Aldrich
Apamin, from bee venom, ≥95% (HPLC)
Sigma-Aldrich
Apamin, synthetic, ≥97% (HPLC)