MilliporeSigma
  • Ultrafine Bi-Sn nanoparticles decorated on carbon aerogels for electrochemical simultaneous determination of dopamine (neurotransmitter) and clozapine (antipsychotic drug).
Ultrafine Bi-Sn nanoparticles decorated on carbon aerogels for electrochemical simultaneous determination of dopamine (neurotransmitter) and clozapine (antipsychotic drug).
Nanoscale (2020-11-04)
Pitchaimani Veerakumar, Shaktivel Manavalan, Shen-Ming Chen, Alagarsamy Pandikumar, King-Chuen Lin
ABSTRACT
This present study describes the synthesis of ultrafine Bi-Sn nanoparticles decorated on carbon aerogels (Bi-Sn NP/CAG) as a nanocomposite for the electrochemical simultaneous determination of dopamine (DA) and clozapine (CLZ). The typical characterization techniques, such as XRD, Raman, BET, FT-IR, TGA, XPS, and FE-SEM/TEM, showed useful insights into the crystal phase and morphology of Bi-Sn NP/CAG. Integrated Bi-Sn NP/CAG built into a cost-effective screen printed carbon electrode (SPCE) offers a high electrochemical surface area (ECSA) compared to unmodified, Bi-Sn, and CAG/SPCEs, such that it favourably allowed the binding of DA and CLZ molecules onto the surface at the Bi-Sn/CAG, which was demonstrated by cyclic and differential pulse voltammetry techniques. As a result, the DA and CLZ sensing exhibited low detection limits (DL, 4.6 and 97.6 nM (S/N = 3)), and sensitivity (3.402 and 0.4 μA μM-1 cm-2) over a wide linear range (0.02-97.59 and 0.5-2092 μM), respectively. To go a step further, the Bi-Sn NP/CAG/SPCE was applied for the simultaneous determination of DA and CLZ which featured lower DL (23.1 and 31.3 nM (S/N = 3)), and sensitivity (0.4979 and 0.04 μA μM-1 cm-2) over a wide linear range (2-182 and 10-910 μM), respectively. The selectivity for DA and CLZ in the presence of a 10-fold concentration of their potentially interfering active species was demonstrated. Finally, this sensing methodology enables the rapid electrochemical determination of the amount of DA and CLZ in a rat brain region serum sample with successful recovery outcomes.
Sigma-Aldrich
Tin(IV) chloride pentahydrate, 98%