MilliporeSigma
  • Oral treatments with a flavonoid-enriched fraction from Cecropia hololeuca and with rutin reduce articular pain and inflammation in murine zymosan-induced arthritis.
Oral treatments with a flavonoid-enriched fraction from Cecropia hololeuca and with rutin reduce articular pain and inflammation in murine zymosan-induced arthritis.
Journal of ethnopharmacology (2020-04-09)
Felipe Marques Teixeira, Mariana Neubarth Coelho, Fernanda do Nascimento José-Chagas, David do Carmo Malvar, Alexandre Kanashiro, Fernando Queiroz Cunha, Marcelo Dias Machado Vianna-Filho, Angelo da Cunha Pinto, Frederico Argollo Vanderlinde, Sônia Soares Costa
ABSTRACT
Cecropia Loefl. species (Urticaceae) are widely spread across the rainforest in tropical and subtropical regions of Central and South America. Inhabitants of different regions of Brazil employ leaves, fruits and sprouts of Cecropia hololeuca Miq. mainly as anti-inflammatory, anti-asthmatic, expectorant, fever suppressant, and against cough. To evaluate the antinociceptive and anti-inflammatory activities of an aqueous leaf extract of C. hololeuca in a murine model of zymosan-induced arthritis (ZIA) and characterize compounds contributing to these effects. The crude aqueous extract of C. hololeuca (CAE) was obtained by infusion, screened for antinociceptive and anti-inflammatory activities, and fractionated (solvent partition; RP-2 and Sephadex G-25 column chromatography), yielding fractions that were chemically and pharmacologically investigated. TLC, HPLC-DAD, HPLC-DAD-ESI-MS/MS and NMR analyses were peformed. The antinociceptive activity was assessed by means of acetic acid-induced writhing, hot-plate and rota-rod tests. ZIA was used to evaluate the anti-arthritic activity of oral treatment with CAE, butanolic (BF) and aqueous fraction (AF), as well as the fractions obtained from BF (F2, F2-A and F2-B). Rutin, a flavonoid found in C. hololeuca, was also tested. Mechanical hypernociception, joint edema, local neutrophil recruitment and articular TNF-α quantification were performed to measure the severity of arthritis and identify the anti-inflammatory potential of C. hololeuca. CAE (0.03-1 g/kg, p.o.) showed a dose-related inhibitory effect on acetic acid-induced writhing test, but did not change the pain latency in the hotplate test, nor the first fall time on the rota-rod test. In addition, CAE (1 g/kg, p.o.) inhibited by 65% the mechanical hypernociception, 46% the joint edema, 54% the neutrophil recruitment and 53% the articular TNF-α concentration levels in ZIA. BF (0.4 g/kg, p.o.), AF (0.6 g/kg), F2 (0.1 g/kg) and F2-A (0.045 g/kg), but not F2-B (0.055 g/kg), inhibited the mechanical hypernociception, joint edema and neutrophil recruitment in ZIA. Rutin (0.001-0.03 g/kg, p.o.) produced dose-related inhibitory effects in the mechanical hypernociception, joint edema and neutrophil recruitment, and at 0.03 g/kg also inhibited articular TNF-α synthesis after intra-articular zymosan injection. Isoorientin, isovitexin, rutin and isoquercitrin were identified in the most active fraction (F2-A), along with luteolin and apigenin derivatives, tentatively identified as isoorientin-2″-O-glucoside and isovitexin-2″-O-glucoside. This study corroborates the popular use by oral route of aqueous preparations of C. hololeuca against joint inflammatory disorders, such as rheumatoid arthritis. Our results demonstrated for the first time that oral administration of rutin shows antinociceptive and anti-inflammatory effects in ZIA, indicating that this flavonoid is one of the immunomodulatory compounds involved in the anti-arthritic activity of C. hololeuca.
Product Description
Rutin, phyproof® Reference Substance