MilliporeSigma
  • Field pea protein isolate/chitosan complex coacervates: Formation and characterization.
Field pea protein isolate/chitosan complex coacervates: Formation and characterization.
Carbohydrate polymers (2020-10-15)
Qing Zhang, Hongmin Dong, Jun Gao, Lingyun Chen, Thava Vasanthan
ABSTRACT
Influence of chitosan (Ch) with low, medium, and high molecular weight (LMW, MMW, and HMW) on the formation of field pea protein isolate (FPPI)/Ch complex coacervates was investigated. An increase in maximum turbidity and a gradual shift of critical pH values towards the isoelectronic point of FPPI were observed as the FPPI/Ch ratio increased. Formation of FPPI/Ch complex coacervates was dominated by the electrostatic and hydrophobic interactions. FPPI/Ch complex coacervates exhibited a porous network microstructure and relatively uniform-sized and even-distributed pores were found in FPPI/Ch-HMW coacervates. Different thermodynamic profiles were observed during complex coacervation between FPPI and Ch with varying MWs and the largest binding stoichiometry was observed in the Ch-MMW at pH 6.6. In summary, the Ch-HMW was demonstrated to be most suitable for the formation of FPPI/Ch complex coacervates with homogenous microstructure but caused less changes in the tertiary conformation of FPPI compared to the Ch-LWM and Ch-MMW.