MilliporeSigma
  • Characterization and antimicrobial evaluation of a new Spgly-AMP, glycine-rich antimicrobial peptide from the mud crab Scylla paramamosain.
Characterization and antimicrobial evaluation of a new Spgly-AMP, glycine-rich antimicrobial peptide from the mud crab Scylla paramamosain.
Fish & shellfish immunology (2020-08-11)
Yichao Xie, Haifu Wan, Xianyuan Zeng, Ziping Zhang, Yilei Wang
ABSTRACT
Antimicrobial peptide (AMP) is a crucial component of the innate immune system in crustaceans. In mud crab, Scylla paramamosain, a commercially important species, a glycine-rich antimicrobial peptide (Spgly-AMP) gene was newly identified and putatively encoded a 26aa signal peptide and 37aa mature peptide. To understand the function of Spgly-AMP, the expression profile of Spgly-amp gene was characterized, which showed Spgly-amp was expressed widely in most tissues of adult crabs with the highest expression level in hemocytes. After Vibrio parahaemolyticus, PGN, or Poly I:C stimulations, the expression level of Spgly-amp was significantly up-regulated in the hemocytes. In antimicrobial assays, chemically synthesized Spgly-AMP peptides exhibited strong antibacterial activities against both Gram-positive and Gram-negative bacteria and high thermal stability after high-temperature heating. These findings in the present study verified the importance of the Spgly-AMP in defense of pathogenic bacteria infection in the mud crab and provided a promising candidate of antimicrobial agents in the crab aquaculture.
Sigma-Aldrich
Peptidoglycan from Bacillus subtilis
Sigma-Aldrich
Polyinosinic–polycytidylic acid potassium salt, with buffer salts, TLR ligand tested