MilliporeSigma
  • Cysteine protease RD21A regulated by E3 ligase SINAT4 is required for drought-induced resistance to Pseudomonas syringae in Arabidopsis.
Cysteine protease RD21A regulated by E3 ligase SINAT4 is required for drought-induced resistance to Pseudomonas syringae in Arabidopsis.
Journal of experimental botany (2020-05-27)
Yi Liu, Kunru Wang, Qiang Cheng, Danyu Kong, Xunzhong Zhang, Zhibo Wang, Qian Wang, Qi Xie, Jijun Yan, Jinfang Chu, Hong-Qing Ling, Qi Li, Jiamin Miao, Bingyu Zhao
ABSTRACT
Plants can be simultaneously exposed to multiple stresses. The interplay of abiotic and biotic stresses may result in synergistic or antagonistic effects on plant development and health. Temporary drought stress can stimulate plant immunity; however, the molecular mechanism of drought-induced immunity is largely unknown. In this study, we demonstrate that cysteine protease RD21A is required for drought-induced immunity. Temporarily drought-treated wild-type Arabidopsis plants became more sensitive to the bacterial pathogen-associated molecular pattern flg22, triggering stomatal closure, which resulted in increased resistance to Pseudomonas syringae pv. tomato DC3000 (Pst-DC3000). Knocking out rd21a inhibited flg22-triggered stomatal closure and compromised the drought-induced immunity. Ubiquitin E3 ligase SINAT4 interacted with RD21A and promoted its degradation in vivo. The overexpression of SINAT4 also consistently compromised the drought-induced immunity to Pst-DC3000. A bacterial type III effector, AvrRxo1, interacted with both SINAT4 and RD21A, enhancing SINAT4 activity and promoting the degradation of RD21A in vivo. Therefore, RD21A could be a positive regulator of drought-induced immunity, which could be targeted by pathogen virulence effectors during pathogenesis.
Sigma-Aldrich
UBE1 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Z-Leu-Leu-Norvalinal, ≥90% (HPLC), powder