MilliporeSigma
  • Selective Partial Hydrolysis of 2-isopropyl-2-oxazoline Copolymers towards Decreasing the Ability to Crystallize.
Selective Partial Hydrolysis of 2-isopropyl-2-oxazoline Copolymers towards Decreasing the Ability to Crystallize.
Materials (Basel, Switzerland) (2020-08-06)
Natalia Oleszko-Torbus, Barbara Mendrek, Agnieszka Kowalczuk, Alicja Utrata-Wesołek, Andrzej Dworak, Wojciech Wałach
ABSTRACT
Poly(2-isopropyl-2-oxazoline) (PiPrOx) is readily prone to crystallization both in solid and from solutions. This feature is detrimental for certain applications. Here, we examine whether the presence of unsubstituted ethyleneimine (EI) units, a gradient distributed within a polymer chain composed of 2-isopropyl-2-oxazoline (iPrOx) and 2-methyl-2-oxazoline (MOx) units, decreases the ability to crystallize the copolymer and affects thermal properties compared to the homopolymer of iPrOx. We assumed that the separation of stiff iPrOx units by the more flexible EI will affect the spatial arrangements of the ordered chains, slightly plasticize and, as a result, decrease their ability to crystallize. The selective hydrolysis of gradient iPrOx and 2-methyl-2-oxazoline (MOx) copolymers, carried out under mild conditions, led to iPrOx/MOx/EI copolymers. To the best of our knowledge, the selective hydrolysis of these copolymers has never been carried out before. Their thermal properties and crystallization abilities, both in a solid state and from an aqueous solution, were analyzed. Based on the analysis of polymer charge and cytotoxicity studies, the potential use of the copolymers obtained was indicated in some biological systems.
Sigma-Aldrich
2-Methyl-2-oxazoline, 98%
Sigma-Aldrich
Methyl 4-nitrobenzenesulfonate, 99%