MilliporeSigma
NBO/NRT Two-State Theory of Bond-Shift Spectral Excitation.
Molecules (Basel, Switzerland) (2020-09-10)
Yinchun Jiao, Frank Weinhold
ABSTRACT
We show that natural bond orbital (NBO) and natural resonance theory (NRT) analysis methods provide both optimized Lewis-structural bonding descriptors for ground-state electronic properties as well as suitable building blocks for idealized "diabatic" two-state models of the associated spectroscopic excitations. Specifically, in the framework of single-determinant Hartree-Fock or density functional methods for a resonance-stabilized molecule or supramolecular complex, we employ NBO/NRT descriptors of the ground-state determinant to develop a qualitative picture of the associated charge-transfer excitation that dominates the valence region of the electronic spectrum. We illustrate the procedure for the elementary bond shifts of SN2-type halide exchange reaction as well as the more complex bond shifts in a series of conjugated cyanine dyes. In each case, we show how NBO-based descriptors of resonance-type 3-center, 4-electron (3c/4e) interactions provide simple estimates of spectroscopic excitation energy, bond orders, and other vibronic details of the excited-state PES that anticipate important features of the full multi-configuration description. The deep 3c/4e connections to measurable spectral properties also provide evidence for NBO-based estimates of ground-state donor-acceptor stabilization energies (sometimes criticized as "too large" compared to alternative analysis methods) that are also found to be of proper magnitude to provide useful estimates of excitation energies and structure-dependent spectral shifts.