MilliporeSigma
  • Glycodendron/pyropheophorbide-a (Ppa)-functionalized hyaluronic acid as a nanosystem for tumor photodynamic therapy.
Glycodendron/pyropheophorbide-a (Ppa)-functionalized hyaluronic acid as a nanosystem for tumor photodynamic therapy.
Carbohydrate polymers (2020-08-25)
Xiaoqin Zhang, Yahui Wu, Zhiqian Li, Wenjia Wang, Yaping Wu, Dayi Pan, Zhongwei Gu, Ruilong Sheng, Helena Tomás, Hu Zhang, João Rodrigues, Qiyong Gong, Kui Luo
ABSTRACT
To enhance the drug delivery efficiency of hyaluronic acid (HA), we designed and prepared glycodendron and pyropheophorbide-a (Ppa)-functionalized HA (HA-Ppa-Dendron) as a nanosystem for cancer photodynamic therapy. Linear Ppa-modified HA (HA-Ppa) was also prepared as a control. Cellular uptake of both polymers by MDA-MB-231 cells led to mitochondrial dysfunction and generation of reactive oxygen species under the irradiation of a laser. Compared to the linear polymer, HA-Ppa-Dendron had higher molecular weight, a more compact nanoscale particle size, and a dendritic structure, resulting in a much longer blood circulation time and higher tumor accumulation. HA-Ppa-Dendron outperformed HA-Ppa in inhibiting cell growth, with 60 % of tumors was eradicated under laser irradiation. Tumor growth inhibition (TGI) up to 99.2 % was achieved from HA-Ppa-Dendron, which was much higher than that of HA-Ppa (50.6 %). Therefore, glycodendron-functionalized HAs by integration of HA and dendritic polymers may act as efficient anti-cancer nanomedicine.
Sigma-Aldrich
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium tetrafluoroborate, 97%