MilliporeSigma
  • Anion-exchanged and quaternary ammonium functionalized MIL-101-Cr metal-organic framework (MOF) for ReO4-/TcO4- sequestration from groundwater.
Anion-exchanged and quaternary ammonium functionalized MIL-101-Cr metal-organic framework (MOF) for ReO4-/TcO4- sequestration from groundwater.
Journal of environmental radioactivity (2020-08-11)
Dien Li, Natalia B Shustova, Corey R Martin, Kathryn Taylor-Pashow, John C Seaman, Daniel I Kaplan, Jake W Amoroso, Roman Chernikov
ABSTRACT
There are few effective technologies for the sequestration of highly water-soluble pertechnetate (TcO4-) from contaminated water despite the urgency of environmental and public health concerns. In this work, anion exchanged and cetyltrimethylammonium bromide (CTAB) functionalized MIL-101-Cr-NO3 were investigated for perrhenate (ReO4-), a surrogate of TcO4-, sequestration from artificial groundwater. Cl-, I-, and CF3SO3- exchanged MIL-101-Cr proved more effective at ReO4- removal than the parent MIL-101-Cr-F. Compared to the parent framework, CTAB functionalized MIL-101-Cr-NO3 increased ReO4- removal capacity from 39 to 139 mg/g, improved the reaction kinetics from ~30 to <10 min to reach full adsorption capacity and the selectivity for ReO4- over competing NO3-, CO32-, SO42-, and Cl-. Spectroscopic data indicated that the chemical speciation of Re in the exchanged MIL-101-Cr remained ReO4-, indicating synergistic sequestration through both anion exchange and non-ion exchange binding with the positively charged ligand of CTAB. These studies foreshadow potential applications of MOFs for the remediation of 99TcO4- from contaminated environments.
Sigma-Aldrich
Aluminum iodide, technical grade, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium perrhenate, 99.99% trace metals basis
Pricing and availability is not currently available.