MilliporeSigma
  • Genomic Methods and Microbiological Technologies for Profiling Novel and Extreme Environments for the Extreme Microbiome Project (XMP).
Genomic Methods and Microbiological Technologies for Profiling Novel and Extreme Environments for the Extreme Microbiome Project (XMP).
Journal of biomolecular techniques : JBT (2017-03-25)
Scott Tighe, Ebrahim Afshinnekoo, Tara M Rock, Ken McGrath, Noah Alexander, Alexa McIntyre, Sofia Ahsanuddin, Daniela Bezdan, Stefan J Green, Samantha Joye, Sarah Stewart Johnson, Don A Baldwin, Nathan Bivens, Nadim Ajami, Joseph R Carmical, Ian Charold Herriott, Rita Colwell, Mohamed Donia, Jonathan Foox, Nick Greenfield, Tim Hunter, Jessica Hoffman, Joshua Hyman, Ellen Jorgensen, Diana Krawczyk, Jodie Lee, Shawn Levy, Natàlia Garcia-Reyero, Matthew Settles, Kelley Thomas, Felipe Gómez, Lynn Schriml, Nikos Kyrpides, Elena Zaikova, Jon Penterman, Christopher E Mason
ABSTRACT
The Extreme Microbiome Project (XMP) is a project launched by the Association of Biomolecular Resource Facilities Metagenomics Research Group (ABRF MGRG) that focuses on whole genome shotgun sequencing of extreme and unique environments using a wide variety of biomolecular techniques. The goals are multifaceted, including development and refinement of new techniques for the following: 1) the detection and characterization of novel microbes, 2) the evaluation of nucleic acid techniques for extremophilic samples, and 3) the identification and implementation of the appropriate bioinformatics pipelines. Here, we highlight the different ongoing projects that we have been working on, as well as details on the various methods we use to characterize the microbiome and metagenome of these complex samples. In particular, we present data of a novel multienzyme extraction protocol that we developed, called Polyzyme or MetaPolyZyme. Presently, the XMP is characterizing sample sites around the world with the intent of discovering new species, genes, and gene clusters. Once a project site is complete, the resulting data will be publically available. Sites include Lake Hillier in Western Australia, the "Door to Hell" crater in Turkmenistan, deep ocean brine lakes of the Gulf of Mexico, deep ocean sediments from Greenland, permafrost tunnels in Alaska, ancient microbial biofilms from Antarctica, Blue Lagoon Iceland, Ethiopian toxic hot springs, and the acidic hypersaline ponds in Western Australia.
Sigma-Aldrich
MetaPolyzyme, lyophilized powder
Pricing and availability is not currently available.