MilliporeSigma
  • Colocalization of MID1IP1 and c-Myc is Critically Involved in Liver Cancer Growth via Regulation of Ribosomal Protein L5 and L11 and CNOT2.
Colocalization of MID1IP1 and c-Myc is Critically Involved in Liver Cancer Growth via Regulation of Ribosomal Protein L5 and L11 and CNOT2.
Cells (2020-04-23)
Ji Hoon Jung, Hyo-Jung Lee, Ju-Ha Kim, Deok Yong Sim, Eunji Im, Sinae Kim, Suhwan Chang, Sung-Hoon Kim
ABSTRACT
Though midline1 interacting protein 1 (MID1IP1) was known as one of the glucose-responsive genes regulated by carbohydrate response element binding protein (ChREBP), the underlying mechanisms for its oncogenic role were never explored. Thus, in the present study, the underlying molecular mechanism of MID1P1 was elucidated mainly in HepG2 and Huh7 hepatocellular carcinoma cells (HCCs). MID1IP1 was highly expressed in HepG2, Huh7, SK-Hep1, PLC/PRF5, and immortalized hepatocyte LX-2 cells more than in normal hepatocyte AML-12 cells. MID1IP1 depletion reduced the viability and the number of colonies and also increased sub G1 population and the number of TUNEL-positive cells in HepG2 and Huh7 cells. Consistently, MID1IP1 depletion attenuated pro-poly (ADP-ribose) polymerase (pro-PARP), c-Myc and activated p21, while MID1IP1 overexpression activated c-Myc and reduced p21. Furthermore, MID1IP1 depletion synergistically attenuated c-Myc stability in HepG2 and Huh7 cells. Of note, MID1IP1 depletion upregulated the expression of ribosomal protein L5 or L11, while loss of L5 or L11 rescued c-Myc in MID1IP1 depleted HepG2 and Huh7 cells. Interestingly, tissue array showed that the overexpression of MID1IP1 was colocalized with c-Myc in human HCC tissues, which was verified in HepG2 and Huh7 cells by Immunofluorescence. Notably, depletion of CCR4-NOT2 (CNOT2) with adipogenic activity enhanced the antitumor effect of MID1IP1 depletion to reduce c-Myc, procaspase 3 and pro-PARP in HepG2, Huh7 and HCT116 cells. Overall, these findings provide novel insight that MID1IP1 promotes the growth of liver cancer via colocalization with c-Myc mediated by ribosomal proteins L5 and L11 and CNOT2 as a potent oncogenic molecule.
Sigma-Aldrich
LX-2 Human Hepatic Stellate Cell Line, a highly suitable model of human hepatic fibrosis
Sigma-Aldrich
MISSION® esiRNA, targeting human CNOT2
Sigma-Aldrich
MISSION® esiRNA, targeting human MID1IP1