MilliporeSigma
  • Transcriptome analysis of Botrytis cinerea in response to tea tree oil and its two characteristic components.
Transcriptome analysis of Botrytis cinerea in response to tea tree oil and its two characteristic components.
Applied microbiology and biotechnology (2020-01-26)
Zhenbiao Li, Xingfeng Shao, Yingying Wei, Kun Dai, Jiayu Xu, Feng Xu, Hongfei Wang
ABSTRACT
Tea tree oil (TTO) and its two characteristic components (terpinen-4-ol and 1,8-cineole) have been shown to inhibit Botrytis cinerea growth. In this study, we conducted a transcriptome analysis to determine the effects of TTO and its characteristic components, alone and in combination, against B. cinerea. Most differentially expressed genes (DEGs) from B. cinerea cells treated with terpinen-4-ol participated in the biosynthesis of secondary metabolites, and the metabolism of amino acids, carbohydrates, and lipids. All treatments containing terpinen-4-ol potentially induced mitochondrial dysfunction and oxidative stress. These were further confirmed by the decreased activities of several enzymes (e.g., succinate dehydrogenase (SDH), malate dehydrogenase (MDH), α-ketoglutarate dehydrogenase (α-KGDH), isocitrate dehydrogenase (ICDH)), the increased activities of certain enzymes (e.g., catalase (CAT), peroxidase (POD), superoxide dismutase (SOD)), and increased content of hydrogen peroxide (H2O2). 1,8-Cineole mainly affected DEGs involved in genetic information processing, resulting in cell death. This study provides insight into the molecular mechanism of B. cinerea inhibition by TTO, and explains the synergistic effect of terpinen-4-ol and 1,8-cineole on B. cinerea.
Sigma-Aldrich
(−)-Terpinen-4-ol, ≥95.0% (sum of enantiomers, GC)
Supelco
Pyrimethanil, PESTANAL®, analytical standard